Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCNT1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12334R
BOSSBS-12334REA 725 EUR InStock
BOSSBS-12334R
Anti-GCNT1 Rabbit Polyclonal Antibody
Antikörper
GCNT1 is expressed in a variety of cell types, including lymphocytes and mucin-producing cells. Specifically, GCNT1 expression in leukocytes regulates the synthesis of core 2 O-glycans that carry sialyl-Lewis x (sLex) oligosaccharides, which confer high affinity binding to Selectins. Downregulation of Selectin ligand expression levels has been shown to inhibit tissue infiltration. Therefore, GCNT1 represents a potential drug target for the treatment of inflammatory disorders and other pathologies involving Selectins.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge