Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HFE Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12335R
BOSSBS-12335REA 704 EUR InStock
BOSSBS-12335R
Anti-HFE Rabbit Polyclonal Antibody
Antikörper
The features of hemochromatosis include cirrhosis of the liver, diabetes, hypermelanotic pigmentation of the skin, and heart failure. Since hemochromatosis is a relatively easily treated disorder if diagnosed, this is a form of preventable cancer. The HFE protein, which is defective in hereditary hemo-chromatosis, normally is expressed in crypt enterocytes of the duodenum where it has a unique, predominantly intracellular localization. In placenta, the HFE protein co-localizes with and forms a stable association with the transferrin receptor (TfR), providing a link between the HFE protein and iron transport. Immunocytochemistry shows that the HFE protein and TfR both are expressed in the crypt enterocytes. Western blots show that, as is the case in human placenta, the HFE protein in crypt enterocytes is physically associated with the TfR and with β2-microglobulin. It is proposed that HFE has two mutually exclusive activities in cells: inhibition of uptake or inhibition of release of iron and that the balance between serum transferrin saturation and serum transferrin-receptor concentrations determines which of these functions predominates. The gene which encodes HFE maps to human chromosome 6p21.3.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HFE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge