Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MAEA Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12337R-CY5
BOSSBS-12337R-CY5EA 900 EUR InStock
BOSSBS-12337R-CY5
Anti-MAEA Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Emp is a 396 amino acid ubiquitously expressed adhesion protein. Expressed as 5 alternatively spliced isoforms, Emp contains one CTLH domain and one LisH domain. Emp can form a complex with F-actin, which is involved regulating actin distribution in erythroblasts and macrophages. Considered to assist with cell division and nuclear architecture, Emp is localized with condensed chromatin at prophase, nuclear spindle poles at metaphase and in the contractile ring during telophase and cytokinesis. Although the exact function of Emp is unknown, Emp is suggested to be involvement in erythroblast-macrophage cell attachment, terminal maturation and enucleation of erythroid cells, and inhibiting apoptosis of erythroblasts.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAEA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge