Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CPA3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12338R-FITC
BOSSBS-12338R-FITCEA 900 EUR InStock
BOSSBS-12338R-FITC
Anti-CPA3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Carboxypeptidase A (CPA) is a pancreatic exopeptidase which hydrolyses the peptide bond adjacent to the C-terminal end in polypeptide chains. Mast cell carboxypeptidase A (MC-CPA), a part of the peptidase M14 family, is a highly conserved metalloprotease localized to the secretory granules, along with trytases and chymases. MC-CPA is stored as an active enzyme in the granule and is released, along with other inflammatory mediators, upon mast cell degranulation. MC-CPA mirrors pancreatic carboxypeptidase A in cleaving COOH-terminal aromatic and aliphatic amino acid residues. The optimum pH of MC-CPA is between neutral and basic, depending upon the substrate. The MC-CPA gene, CPA3, resides on chromosome 3 and contains 11 exons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPA3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge