Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VNN2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12340R-HRP
BOSSBS-12340R-HRPEA 900 EUR InStock
BOSSBS-12340R-HRP
Anti-VNN2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Vanin-2 is a 520 amino acid GPI-anchor protein that belongs to the CN hydrolase family and BTD/VNN subfamily.Widely expressed with high expression in spleen and blood, vanin-2 is suggested to be involved in thymus homing of bone marrow cells, regulation of Integrin ∫2-mediated cell adhesion, and migration and motility of neutrophils. Vanin-2 exists as five alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 6q23.2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: VNN2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge