Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-L3MBTL3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12342R-CY5
BOSSBS-12342R-CY5EA 900 EUR InStock
BOSSBS-12342R-CY5
Anti-L3MBTL3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Polycomb group (PcG) proteins are important for maintaining the transcriptionally repressed state of target genes and are thought to function via chromatin modification. PcG proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. PcG proteins are also required for normal maturation of myeloid progenitor cells. A putative PcG protein, L3MBTL3 (lethal(3)malignant brain tumor-like protein 3), also known as MBT-1 or KIAA1798, is a 780 amino acid protein containing three MBT repeats and one SAM (sterile alpha motif) domain. Localized to the nucleus, L3MBTL3 interacts with RING1B, another PcG protein that may be involved in the specification of anterior-posterior axis and cell proliferation in early development. L3MBTL3 exists as two isoforms produced by alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: L3MBTL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge