Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12343R-FITC
BOSSBS-12343R-FITCEA 900 EUR InStock
BOSSBS-12343R-FITC
Anti-TRIB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
TRB-1 is a 372 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase superfamily. Expressed ubiquitously with highest expression in bone marrow, thyroid gland, skeletal muscle and pancreas, TRB-1 interacts with MAPK kinases and is thought to regulate the activation of MAP kinses, possibly controlling MAP kinase cascades. The gene encoding TRB-1 maps to human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRIB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge