Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EDA Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12347R-CY3
BOSSBS-12347R-CY3EA 909 EUR InStock
BOSSBS-12347R-CY3
Anti-EDA Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Affected males of X-linked anhidrotic ectodermal dysplasia show hypotrichosis, abnormal teeth and absent sweat glands. Some of the patients reported by Halperin and Curtis showed mental defect also, but this is not an invariable feature. Ectodysplasin A (EDA) is a trimeric type II membrane protein that co-localizes with cytoskeletal structures at the lateral and apical surfaces of cells. EDA is expressed in hair follicles and in the epidermis of adult skin. The sequence of the longest isoform includes an interrupted collagenous domain of 19 Gly-X-Y repeats and a motif conserved in the tumor necrosis factor (TNF)-related ligand family. EDA is a member of the TNF-related ligand family involved in the early epithelial-mesenchymal interaction that regulates ectodermal appendage formation. Similar to other members of collagenous membrane proteins and members of TNF-related ligands, EDA is a type II membrane protein which forms trimers.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge