Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MIXL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12350R-FITC
BOSSBS-12350R-FITCEA 900 EUR InStock
BOSSBS-12350R-FITC
Anti-MIXL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and in the control of gene expression. MIXL1 (Mix1 homeobox-like 1), also known as MIXL, is a 232 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. Expressed in lymph tissues, MIXL1 functions as a transcription factor that plays an essential role in axial mesendoderm morphogenesis and endoderm formation and is also required for cellular differentiation during blood development. Additionally, MIXL1 is involved in maturation of heart and gut tissue during embryogenesis and may also act as a negative regulator of brachyury expression. Overexpression of MIXL1 is associated with non-Hodgkin and Hodgkin lymphomas, suggesting a role in carcinogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MIXL1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge