Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-STRA6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12351R-FITC
BOSSBS-12351R-FITCEA 900 EUR InStock
BOSSBS-12351R-FITC
Anti-STRA6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
STRA6 is a 667 amino acid, multi-pass cell membrane protein. Stra6 functions as a cell-surface receptor for the complex retinol-retinol binding protein (RBP/RBP4). Ultimately increasing cellular retinol uptake from the retinol-RBP complex, Stra6 removes retinol from RBP/RPB4 and transports it across the plasma membrane, where it is metabolized. Stra6 is broadly expressed, with 4 named isoforms that exist as a result of alternative splicing events. Mutations in the gene encoding Stra6 cause Matthew-Wood Syndrome, also known as Spear Syndrome. This syndrome is characterized by anophtalmia, mild facial dysmorphism and malformations of the heart, lung and diaphragm. The Stra6 gene maps to chromosome 15q24.1.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: STRA6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge