Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HES7 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12353R-CY5
BOSSBS-12353R-CY5EA 900 EUR InStock
BOSSBS-12353R-CY5
Anti-HES7 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
HES7 is a 225 amino acid transcriptional repressor protein. Localized to the nucleus, HES7 represses transcription of N box- and E box-containing promoters. HES7, along with family member HES1, is thought to cooperatively regulate somite formation in the presomitic mesoderm. HES7 may also be essential for coordinated somite segmentation by acting as a segmentation clock. HES7 contains one basic helix-loop-helix (bHLH) domain and one Orange domain. Mutations in HES7 have been found to cause spondylocostal dysostosis, an autosomal recessive disorder characterized by deformities of the chest and spine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HES7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge