Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ASPSCR1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12355R-CY3
BOSSBS-12355R-CY3EA 900 EUR InStock
BOSSBS-12355R-CY3
Anti-ASPSCR1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Glut4 is a twelve pass transmembrane protein (12TM) whose carboxy-terminus may dictate its cellular localization. Aberrant Glut4 expression has been suggested to contribute to such maladies as obesity and diabetes. Glut4 null mice have shown that while functional Glut4 protein is not required for maintaining normal glucose levels, it is necessary for sustained growth, normal cellular glucose, fat metabolism and prolonged longevity. TUG (ASPL in humans) regulates the trafficking of glucose via Glut4. Full-length TUG forms a complex with Glut4 and in 3T3-L1 adipocytes and this complex is present in unstimulated cells and is disassembled by insulin. TUG acts by trapping endocytosed Glut4 and tethering it intracellularly. Insulin mobilizes this pool of retained Glut4 by releasing the tether.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASPSCR1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge