Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ALX1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12356R-A647
BOSSBS-12356R-A647EA 893 EUR InStock
BOSSBS-12356R-A647
Anti-ALX1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
ALX1 is a 326 amino acid protein specific to cervix and cartilage tissues. As well as having a homeobox domain, ALX1 also contains an OAR domain, which has been suggested to be important for DNA binding or protein-protein interactions and transactivation. First characterized from a rat chondrosarcoma tumor cell line, ALX1 is a homeobox transcription factor that regulates downstream target genes and has specifically shown to act as a transcriptional repressor for rat prolactin in vivo. Homozygous ALX1 deficient mice are born with acrania and meroanencephaly, suggesting ALX1 function in the development of the neural tube. It has also been suggested that ALX1 cooperates with ALX4 in limb development and craniofacial bone formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge