Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CFDP1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12357R
BOSSBS-12357REA 725 EUR InStock
BOSSBS-12357R
Anti-CFDP1 Rabbit Polyclonal Antibody
Antikörper
CFDP1 is a 299 amino acid protein that is involved in embryogenesis and normal cell function. When treated with CFDP1 peptide, mouse molar teeth increase in size, whereas treating cells with against CFDP1 shows an increase in the number of apoptotic cells and gradual tooth disintegration. CFDP1 is highly expressed in developing mouse teeth and is expressed at lower levels in liver, lung and heart. The gene encoding CFDP1 maps to human chromsome 16, in a region that has been associated with inherited craniofacial diseases, such as fanconi anemia type A. There are two isoforms of CFDP1 that are produced as a result of alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CFDP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge