Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-YKL39/CHI3L2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12358R-A750
BOSSBS-12358R-A750EA 890 EUR InStock
BOSSBS-12358R-A750
Anti-YKL39/CHI3L2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
YKL-39 is a 390 amino acid cartilage protein that belongs to the chitinase family of chitin-fragmenting hydrolases. Highly expressed in chondrocytes (cartilage cells) and synoviocytes (fibroblastic cells that line joint cavities), YKL-39 binds glycan structures with high affinity. Although related to bacterial chitinases, YKL-39 lacks the characteristic glutamate active site and, thus, does not have enzymatic chitinase activity. Patients affected with rheumatoid arthritis (RA) have autoimmunity against YKL-39, suggesting that YKL-39 is involved in osteoarthritic and/or rheumatoid joint disease. Additionally, YKL-39 is upregulated in early degenerative cartilage diseases (such as RA) and may be a marker of chondrocyte activation in these autoimmune conditions.

Type: Primary
Antigen: YKL39/CHI3L2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-220/390
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge