Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DMP1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12359R-CY5
BOSSBS-12359R-CY5EA 883 EUR InStock
BOSSBS-12359R-CY5
Anti-DMP1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DMP-1 is a member of the small integrin ligand N-linked glycoprotein family. It is important for the mineralization of bone and dentin. DMP-1 is expressed in bone, tooth and hypertrophic cartilage. It is synthesized by preosteoblasts and contains a large number of acidic domains. DMP-1 localizes to the nucleus of undifferentiated osteoblasts where it functions as a transcriptional regulator for osteoblast-specific gene activation and induces osteoblast differentiation. During osteoblast maturation, DMP-1 undergoes a conformational change and becomes phosphorylated by casein kinase II in response to an influx of calcium ions to the nucleus. DMP-1 is then exported to the extracellular matrix (ECM) where it regulates the nucleation of hydroxyapatite and the formation of calcified tissue. DMP-1 is proteolytically processed into N- and C-terminal fragments in the ECM of bone and dentin. The protein has also been identified in bone as a high molecular weight proteoglycan comprised of the N-terminal DMP-1 fragment and chondroitin sulfate.The loss of DMP-1 can result in hypomineralized bone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DMP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge