Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SUMF1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12366R-CY3
BOSSBS-12366R-CY3EA 900 EUR InStock
BOSSBS-12366R-CY3
Anti-SUMF1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
SUMF1 is a 374 amino acid alternatively spliced protein that localizes to the lumen of the endoplasmic reticulum and belongs to the sulfatase-modifying factor family. Expressed ubiquitously with highest expression in liver, kidney and pancreas, SUMF1 exists as either a monomer, a homodimer or a heterodimer (with SUMF2) and functions to oxidize sulfatase cysteine residues to an active FGIy residue, thereby playing an important role in sulfatase activity. Defects in the gene encoding SUMF1 are the cause of multiple sulfatase deficiency (MSD), a heterogeneous disorder characterized by metachromatic leukodystrophy, mucopolysaccharidosis, chondrodysplasia punctata, hydrocephalus, ichthyosis, neurologic deterioration and developmental delay.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SUMF1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge