Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MEGF10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12372R-FITC
BOSSBS-12372R-FITCEA 900 EUR InStock
BOSSBS-12372R-FITC
Anti-MEGF10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
MEGF10 is a 1,140 amino acid protein encoded by the human gene MEGF10. MEGF10 belongs to the MEGF family and contains fifteen EGF-like domains and one EMI domain. The engulfment of dying cells is a specialized form of phagocytosis that is highly conserved across evolution and is mediated by the concerted action of several molecules. MEGF10 is an engulfment receptor protein that localizes to the plasma membrane in a punctuated pattern. The MEGF10 protein shares structural similarities with nematode engulfment receptor cell death abnormal-1 (CED-1). MEGF10 function is modulated by the ATP binding cassette transporter ABCA1, which is an ortholog to CED-7. It is expressed at the cell surface in clusters around cell corpses during engulfment, and during the engulfment of apoptotic thymocytes is recruited to the bottom of the forming phagocytic cup.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MEGF10
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge