Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UCMA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12378R-HRP
BOSSBS-12378R-HRPEA 900 EUR InStock
BOSSBS-12378R-HRP
Anti-UCMA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
UCMA is a 138 amino acid secreted protein that is highly expressed in resting chrondrocytes in developing long bones and is thought to function in the early phase of chrondrocyte differentiation. A furin-like protease processes UCMA into an N-terminal 37 amino acid peptide and a C-terminal 74 amino acid peptide, which is referred to as Unique cartilage matrix-associated protein C-terminal fragment (Ucma-C). Introduction of recombinant Ucma-C interferes with osteogenic differentiation of mesenchymal stem cells, MC3T3-E1 preosteoblasts and primary osteoblasts. This suggests that Ucma may be involved in the negative regulation of osteogenic differentiation of osteochondrogenic precursor cells at the cartilage-bone interface and in peripheral zones of fetal cartilage.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: UCMA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge