Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-STC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12379R-A647
BOSSBS-12379R-A647EA 893 EUR InStock
BOSSBS-12379R-A647
Anti-STC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Stanniocalcin 1 (STC1) and stanniocalcin 2 (STC2) are mammalian peptide hormones that were previously considered to be present only in bony fish, where they are involved in calcium homeostasis. STC1 plays a role in calcium and phosphate homoeostasis and is phosphorylated in vitro by protein kinase C, and STC2 is phosphorylated in vitro by casein kinase II (CK2). A human fibrosarcoma cell line, HT1080, expresses both STC1 and STC2 as secreted phosphoproteins in vivo, with STC2 being phosphorylated by an ecto-CK2-like enzyme. STC1 and STC2 have opposite effects on calcium and phosphate homeostasis, namely anti-hypercalcemic and anti-hypocalcemic actions, respectively. STC1 and STC2 are detected in human adrenal tumors, such as pheochromocytoma, differentiated neuroblastoma aldosterone-producing adenoma, and in cultured adrenal tumor cells (rat pheochromocytoma PC-12 cells and human neuroblastoma NB-1 cells).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge