Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EYA3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12382R-A647
BOSSBS-12382R-A647EA 893 EUR InStock
BOSSBS-12382R-A647
Anti-EYA3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
EYA3 is a 573 amino acid protein that localizes to both the nucleus and the cytoplasm and is one of several mammalian homologs of the Drosophila Eya (eyes absent) protein. Existing as two alternatively spliced isoforms, EYA3 possesses magnesium-catalyzed phosphatase activity and is thought to play a role in transcriptional regulation during organogenesis. Specifically, EYA3 interacts with proteins such as Six1 and, via this interaction, functions to activate the expression of genes that are involved in cellular proliferation and organ development. Upon DNA damage, EYA3 may be phosphorylated by ATM or ATR. The gene encoding EYA3 maps to chromosome 1, which spans about 260 million base pairs and comprises nearly 8% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EYA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge