Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MFNG Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12390R-A350
BOSSBS-12390R-A350EA 893 EUR InStock
BOSSBS-12390R-A350
Anti-MFNG Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Three mammalian fringe family members, Manic, Radical and Lunatic Fringe, have been identified as proteins related to Drosophila fringe, a protein involved in development. Fringe proteins act upstream of the Notch signaling pathway and are involved in boundary determination during segmentation. Each mammalian fringe displays different patterns of expression, though all are expressed in the mouse embryo as well as in many adult tissues. Radical fringe plays a key role in the development of the limb bud. Lunatic fringe is required for normal somite segmentation and patterning and is thought to be a target of the molecular clock. Manic fringe, also involved in somatic development, has been shown to render mouse NIH/3T3 cells tumorigenic in SCID mice.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MFNG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge