Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Mib1/Mindbomb Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12391R-A750
BOSSBS-12391R-A750EA 890 EUR InStock
BOSSBS-12391R-A750
Anti-Mib1/Mindbomb Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
MIB1 is a 1006 amino acid E3 ubiquitin ligase that activates the Notch ligand, Delta. MIB1 ubiquinates Delta by binding to its intracellular domain, leading to the endocytosis and eventual degradation of the Delta receptor, which, paradoxically, results in the up-regulation of receptor activity and enhances Notch Signalling. MIB1 also interacts with DAPK, a protein that plays an important role in the regulation of apoptosis. Ubiquination of DAPK leads to inhibition of caspase-dependent apoptosis, therefore it is likely that overexpression of MIB1 can lead to tumour growth. Although it seems to be widely expressed at low levels, MIB1 is expressed at highest concentrations in the CNS and ovary. Both DAPK and MIB1 are overexpressed in epileptic brain tissue, suggesting that they probably cooperate as regulators of neuronal death in epilepsy.

Type: Primary
Antigen: Mib1/Mindbomb
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/1006
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge