Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TLE5/Amino-terminal enhancer of split Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12392R-A680
BOSSBS-12392R-A680EA 890 EUR InStock
BOSSBS-12392R-A680
Anti-TLE5/Amino-terminal enhancer of split Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
TLE5 is a 197 amino acid nuclear protein that belongs to the TLE family. Expressed predominately in fetal brain, liver, lung, heart and kidney and in adult muscle, TLE5 functions as either a homooligomer or a heterooligomer with other TLE family members and, through this association, dominantly represses the expression of TLE genes. In addition, TLE5 can repress NFkB-regulated gene expression and is thought to play an important role in initiating and maintaining cell differentiation events. Two isoforms of TLE5 exist due to alternative splicing events.

Type: Primary
Antigen: TLE5/Amino-terminal enhancer of split
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/197
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge