Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AMHR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12414R-A647
BOSSBS-12414R-A647EA 893 EUR InStock
BOSSBS-12414R-A647
Anti-AMHR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
MISR II is a 573 amino acid protein encoded by the human gene AMHR2. MISR II belongs to the protein kinase superfamily, TKL Ser/Thr protein kinase family, TGFB receptor subfamily and contains one protein kinase domain. Upon ligand binding, MISR II forms a receptor complex consisting of two type II and two type I transmembrane serine/threonine kinases. These type II receptors rylate and activate type I receptors which autophosphorylate, then bind and activate Smad transcriptional regulators. MISR II also acts as a receptor for anti-Muellerian hormone. Defects in AMHR2 are the cause of persistent Muellerian duct syndrome type 2 (PMDS-2). PMDS-2 is a form of male pseudo-hermaphroditism characterized by a failure of Muellerian duct regression in otherwise normal males.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMHR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge