Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACVR2A Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12416R-CY5
BOSSBS-12416R-CY5EA 1350 EUR InStock
BOSSBS-12416R-CY5
Anti-ACVR2A Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Members of the transforming growth factor b superfamily bind to a pair of transmembrane proteins, known as receptor types I and II, which contain serine/threonine kinases and associate to form a signaling complex (1). Activin has been shown to bind a heteromeric noncovalent complex, which consists of a type I receptor, ACTR-IA (also designated ACVRI and ALK-2) or ACTR-IB (also designated ALK-4 and SKR2), and a type II receptor, ACTR-IIA (also designated ACVR2A) or ACTR-IIB (also designated ACVR2B) (1-6). Both receptor types are highly expressed in brain (5). The activin receptor family members are thought to mediate distinct effects on gene expression, cell differentiation, and morpho- genesis in a dose dependent fashion (5,6).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACVR2A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge