Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAPRIN2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12423R
BOSSBS-12423REA 725 EUR InStock
BOSSBS-12423R
Anti-CAPRIN2 Rabbit Polyclonal Antibody
Antikörper
Caprin2 is 1127 amino acid highly conserved protein that is ubiquitously expressed with highest levels of expression in brain and spleen. Caprin2 stabilizes cytosolic β-catenin and enhances LEF-1 dependent reporter gene activity as well as the expression of Wnt target genes in mammalian cells. Caprin2 promotes LRP5/6 phosphorylation by GSK-3 and enhances the interaction between Axin and LRP5/6. It is suggested that Caprin2 functions as a proapoptotic inhibitor of the cell cycle. Nine isoforms of Caprin2 exist due to alternative splicing events.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CAPRIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge