Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRRFIP2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12438R-A555
BOSSBS-12438R-A555EA 893 EUR InStock
BOSSBS-12438R-A555
Anti-LRRFIP2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
LRRFIP2 is a 721 amino acid protein that belongs to the LRRFIP family. Ubiquitously expressed, LRRFIP2 acts as an activator of the Wnt signaling pathway and as a positive regulator of NFâ…¹B activity. LRRFIP2 may be involved in regulating cytokine production in macrophages, suggesting a functional role in the TLR4-mediated inflammatory response. Three isoforms of LRRFIP2 exists due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRFIP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge