Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ABLIM3 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12442R
BOSSBS-12442REA 725 EUR InStock
BOSSBS-12442R
Anti-ABLIM3 Rabbit Polyclonal Antibody
Antikörper
The ABLIM1 protein has an N-terminal domain that contains four double zinc finger motifs, which conform to the LIM motif consensus sequence. ABLIM1 binds to F-Actin through a dematin-like domain and is expressed in retina, brain and muscle tissue. There are four known isoforms of ABLIM1. The gene encoding ABLIM1 maps to a region of chromosome 10 associated with frequent loss of heterozygosity in human tumors, thus identifying ABLIM1 as a candidate tumor suppressor gene. ABLIM2 and ABLIM3 show highest expression in muscle and neuronal tissues, bind to F-Actin, and are localized on stress fibers. They also have been shown to enhance STARS (striated muscle activator of Rho signaling) dependent activation of serum-response factor (SRF), thereby modulating transcription.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ABLIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge