Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADH6 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12449R-CY3
BOSSBS-12449R-CY3EA 900 EUR InStock
BOSSBS-12449R-CY3
Anti-ADH6 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ADH6 (alcohol dehydrogenase 6), also known as ADH-5, is a 368 amino acid member of the class V zinc-containing alcohol dehydrogenase family. This family of enzymes functions to metabolize a wide variety of substrates such as retinol, hydroxysteroids, ethanol, aliphatic alcohols and lipid peroxidation products. Localized to the cytoplasm and expressed in the stomach and liver, ADH6 catalyzes the reversible oxidation of alcohols to their corresponding aldehydes or ketones and is able to bind two zinc ions as cofactors. ADH6 contains a glucocorticoid response element upstream of its 5' UTR which is thought to be a steroid binding site, suggesting that expression of ADH6 may be under hormonal control. Multiple isoforms of ADH6 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADH6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge