Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ALDH9A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12461R-A647
BOSSBS-12461R-A647EA 893 EUR InStock
BOSSBS-12461R-A647
Anti-ALDH9A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH9A1 (aldehyde dehydrogenase family 9 member A1), also known as E3, ALDH4, ALDH7, ALDH9 or TMABADH, is a 494 amino acid cytoplasmic protein that is highly expressed in adult liver, skeletal muscle, kidney and embryonic brain. ALDH9A1 converts gamma-trimethylaminobutyraldehyde into gamma-butyrobetaine and catalyzes the irreversible oxidation of a broad range of aldehydes to the corresponding acids in a NAD-dependent reaction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge