Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AGPS/Alkyl-DHAP synthase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12462R-A750
BOSSBS-12462R-A750EA 890 EUR InStock
BOSSBS-12462R-A750
Anti-AGPS/Alkyl-DHAP synthase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
AGPS is a 658 amino acid enzyme that is required for glycerolipid metabolism and ether lipid biosynthesis. localised to the inner aspect of the peroxisomal membrane, AGPS is likely part of a heterotrimeric complex that is also composed of GNPAT and a modified form of GNPAT. Containing one FAD-binding PCMH-type domain, AGPS utilizes FAD as a cofactor in the synthesis of alkyl-glycerone 3-phophate and a long-chain acid anion from 1-acteyl-glyerone 3-phosphate and a long-chain alcohol. Defects in the gene encoding AGPS results in rhizomelic chondrodysplasia punctata type 3, a disease characterised by vertebral disorders, severe mental retardation, cutaneous lesions, cataracts and rhizomelic shortening of the humerus and femur.

Type: Primary
Antigen: AGPS/Alkyl-DHAP synthase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/658
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge