Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ANAPC11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12484R-A555
BOSSBS-12484R-A555EA 893 EUR InStock
BOSSBS-12484R-A555
Anti-ANAPC11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Comprising more than ten subunits, the anaphase-promoting complex (APC) acts in a cell-cycle dependent manner to promote the separation of sister chromatids during the transition between metaphase and anaphase in mitosis. APC, or cyclosome, accomplishes this progression through the ubiquitination of mitotic cyclins and other regulatory proteins that are targeted for destruction during cell division. APC is phosphorylated, and thus activated, by protein kinases Cdk1/cyclin B and polo-like kinase (Plk). APC is under tight control by a number of regulatory factors, including CDC20, CDH1 and MAD2. Specifically, CDC20 and CDH1 directly bind to APC and activates APC’s cyclin-ubiquitination activity. In contrast, MAD2 inhibits APC by forming a ternary complex with CDC20 and APC; thus preventing APC activation. APC11 is a RING-H2 finger protein that allows for the synthesis of multiubiquitin chains in the presence of Ubiquitin carrier protein 4 (Ubc4) and ubiquitin conjugating enzyme (E2). In addition, a heterodimeric complex of either Ubc4 or UbcH10 with APC11 and APC2 catalyzes the ubiquitination of human securin and cyclin B1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANAPC11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge