Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APLF Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12489R-A680
BOSSBS-12489R-A680EA 890 EUR InStock
BOSSBS-12489R-A680
Anti-APLF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
APLF is a 511 amino acid protein that contains one FHA domaIn and two C2H2type zinc fingers. localised to both the nucleus and the cytoplasm, APLF interacts with XRCC1, XRCC4 and Ku-86 and, via these interactions, is involved in single-strand and double-strand DNA break repair. APLF is subject to post-translational phosphorylation in response to DNA breaks. The gene encoding APLF maps to human chromosome 2, which houses over 1400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.

Type: Primary
Antigen: APLF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-511/511
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge