Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APOL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12499R-HRP
BOSSBS-12499R-HRPEA 900 EUR InStock
BOSSBS-12499R-HRP
Anti-APOL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The apolipoprotein L gene family maps to a region on chromosome 22 and encodes six highly homologous proteins designated apoL-I, apoL-II, apoL-III, apoL-IV, apoL-V and apoL-VI, all of which function as components of plasma lipoproteins. ApoL-IV (apolipoprotein L-IV), also known as APOL4, is a 351 amino acid protein that exists as multiple alternatively spliced isoforms, one of which is secreted. Expressed in spleen, placenta, spinal cord, uterus, testis and trachea, apoL-IV is thought to play a role in lipid exchange and transport throughout the body and may be involved in reverse cholesterol transport, specifically from peripheral cells to the liver. Overexpression of apoL-IV is associated with schizophrenia, suggesting that apoL-IV may play a role in the pathogenesis of neural disorders.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: APOL4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge