Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APOL5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12500R-A350
BOSSBS-12500R-A350EA 893 EUR InStock
BOSSBS-12500R-A350
Anti-APOL5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The apolipoprotein L gene family maps to a region on chromosome 22 and encodes six highly homologous proteins designated apoL-I, apoL-II, apoL-III, apoL-IV, apoL-V and apoL-VI, all of which function as components of plasma lipoproteins. ApoL-V (apolipoprotein L-V), also known as APOL5, is a 433 amino acid protein that localizes to the cytoplasm and belongs to the apolipoprotein L family. Expressed in a variety of tissues including testis, stomach, uterus and skeletal muscle, apoL-V is thought to affect the movement of lipids in the cytoplasm and may allow the binding of lipids to organelles. Like other members of the apolipoprotein L family, apoL-V is thought to be involved in the development of schizophrenia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOL5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge