Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APOO Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12502R-HRP
BOSSBS-12502R-HRPEA 900 EUR InStock
BOSSBS-12502R-HRP
Anti-APOO Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Apolipoproteins are a family of fatty-acid binding proteins that transport fat through the bloodstream in the form of lipoproteins. ApoO (Apolipoprotein O), also known as FAM121B or My025, is a 198 amino acid single-pass membrane protein that belongs to the apolipoprotein family. Expressed ubiquitously with particularly high expression in diabetic heart tissue, apoO functions to promote the transport of cholesterol from macrophage cells and may be involved in regulatory mechanisms that protect lipid accumulation within the heart. ApoO is present in high density lipoproteins (HDLs) and low density lipoproteins (LDLs) and is secreted by an MTP (microsomal triglyceride transfer protein)-dependent mechanism. Two isoforms of apoO exist due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: APOO
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge