Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ASL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12515R-FITC
BOSSBS-12515R-FITCEA 900 EUR InStock
BOSSBS-12515R-FITC
Anti-ASL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ASL is a member of the lyase 1 family of proteins and is predominantly expressed in the liver. Localizing to the cytoplasm and existing as a homotetramer, ASL catalyzes the hydrolytic cleavage of argininosuccinic acid (ASA) to fumarate and arginine, an essential step of the urea cycle which is crucial for the detoxification of ammonia. This reaction is also involved in the biosynthesis of arginine. In addition, ASL shares high sequence homology with the avian and reptilian eye lens protein, d-crystallin. Mutations in the gene encoding ASL leads to an accumulation of ASA in body fluids and results in Arginosuc-cinic aciduria (ASAuria), an autosomal recessive disorder that is characterized by hyperammonemia, liver enlargement, convulsions, physical and mental retardation, episodic unconsciousness and dry and brittle hair showing trich-orrhexis nodosa (weak points or nodes in the hair shaft).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge