Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12522R-A488
BOSSBS-12522R-A488EA 893 EUR InStock
BOSSBS-12522R-A488
Anti-ARL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The ADP-ribosylation factor (ARF) protein family are structurally and functionally conserved members of the Ras superfamily of regulatory GTP-binding proteins (1–3). ARFs influence vesicle trafficking and signal transduction in eukaryotic cells (1–3). ARF-dependent regulatory mechanisms include the coordination of spectrin interactions with golgi membranes and the association of actin to the golgi via rho family-dependent G-protein localization (Rac, CDC42) and WASP/Arp2/3 complexes (3–7). Additionally, ARFs play a central role in maintenance of organelle integrity, assembly of coat proteins, and activation of phospholipase D (5–7). The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6); members of each class share a common gene organization (8,9). The human ARF6 gene maps to chromosome 7q22.1, contains five exons and four introns, and encodes a 175 amino acid protein (8,9).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge