Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATXN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12534R-A647
BOSSBS-12534R-A647EA 921 EUR InStock
BOSSBS-12534R-A647
Anti-ATXN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Ataxin-1, also designated spinocerebellar ataxia type 1 protein (Sca-1), is differentially expressed and localizes to both the cytoplasm and the nucleus. Mutations in Ataxin-1 are associated with the onset of the autosomal dominant neurodegenerative disorder spinocerebellar ataxia type 1 (SCA-1), which is characterized by progressive neuronal loss in the cerebellum, muscle wasting and ataxia. In Purkinje cells, where SCA-1 is predominantly observed, Ataxin-1 has been shown to directly associate with the Purkinje-enriched leucine-rich acidic nuclear protein (LANP) and the nuclear matrix-associated protein promyelocytic leukemia protein PML. In SCA-1, Ataxin-1 is mutated to encode a polyglutamine protein that forms nuclear aggregates, which interact significantly more strongly with LANP and contribute to the pathogenesis of SCA-1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATXN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge