Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PURH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12544R-HRP
BOSSBS-12544R-HRPEA 900 EUR InStock
BOSSBS-12544R-HRP
Anti-PURH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The bifunctional purine biosynthesis protein ATIC (also designated PURH) contains AICAR transformylase and IMP cyclohydrolase activities. AICAR (5-aminoimidazole-4-carboxamide ribonucleotide) transformylase catalyzes the second to last step in purine biosynthesis, playing an important role in the production of nucleotides and IMP. Defects in the ATIC transformylase gene can cause AICA-rebsuria, also designated AICA-ribosiduria, an inborn error in purine biosynthesis that is neurologically cataclysmic. Individuals with AICA-rebosuria accumulate AICA-riboside, also designated ZMP, and its derivatives in erythrocytes and fibroblasts. Patients also excrete very large amounts of AICA-riboside in the urine. Mental retardation, epilepsy, dysmorphic features and congenital blindness are all symptoms of this disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PURH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge