Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATRN Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12551R-CY7
BOSSBS-12551R-CY7EA 900 EUR InStock
BOSSBS-12551R-CY7
Anti-ATRN Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Mahogany (MG), originally identified as a protein involved in pigmentation, acts in conjunction with melanocortin receptors to suppress diet-induced obesity. Mahogany contains a single transmembrane domain, and it is expressed in a broad range of tissues, including the hypothalamus and pigment cells. Mutations within the mahogany gene were shown to rescue agouti-lethal-yellow mutant mice from obesity. The extracellular domain of mouse mahogany is the ortholog of the human protein attractin. Attractin (also designated DPPT-L) is a human serum glycoprotein and is a member of the CUB family of cell adhesion and guidance proteins. Attractin is expressed on activated T cells and is released from the cells 48 to 72 hours after activation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge