Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ALKBH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12565R-A488
BOSSBS-12565R-A488EA 893 EUR InStock
BOSSBS-12565R-A488
Anti-ALKBH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ALKB protects DNA and RNA against damage from methylating compounds from the environment by directly reversing 1-methyladenine (1-meA) and 3-methylcytosine (3-meC) cytotoxic alkylation lesions in DNA and RNA. The enzymes act by oxidative demethylation, utilizing ferrous iron and alpha-ketoglutarate as cofactors, 2-oxoglutarate as a co-substrate, and molecular oxygen as the oxidizing agent. Deficiencies in DNA and RNA repair in mammals are associated with cancer, neurological disease and developmental defects. ALKB plays a role in resistance to anti-cancer drugs which attempt to damage tumor DNA. Escherichia coli ALKB protein belongs to the superfamily of 2-oxoglutarate- and iron(II)-dependent oxygenases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALKBH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge