Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BDP/ARID3B Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12580R-A750
BOSSBS-12580R-A750EA 890 EUR InStock
BOSSBS-12580R-A750
Anti-BDP/ARID3B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ARID3B represents a member of a unique family of DNA-binding proteins that have roles in cell lineage gene regulation, cell cycle control, embryonic patterning and transcriptional regulation. ARID3B localizes to the nucleus and contains an AT-rich interactive domain (ARID) which has DNA-binding capabilities. Functioning as a transcription factor, ARID3B associates in a heterodimer with the related protein ARID3A and is believed to play an important role in neural crest survival during embryogenesis. In addition, ARID3B may participate in malignant transformation and neuroblastoma growth, suggesting a possible use of ARID3B as a tumour marker for neuroblastoma.

Type: Primary
Antigen: BDP/ARID3B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/561
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge