Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRYBB1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12582R
BOSSBS-12582REA 725 EUR InStock
BOSSBS-12582R
Anti-CRYBB1 Rabbit Polyclonal Antibody
Antikörper
Crystallins are the major proteins of the vertebrate eye lens, where they maintain the transparency and refractive index of the lens. Crystallins are divided into Alpha, Beta, and Gamma families, and the Beta- and Gamma-crystallins also comprise a superfamily. Crystallins usually contain seven distinctive protein regions, including four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins constitute the major lens structural proteins, and they associate into dimers, tetramers, and higher order aggregates. The Beta-crystallin subfamily is composed of several gene products, including Beta A1-, Beta A2-, Beta A3-, Beta A4-, Beta B1-, Beta B2- and Beta B3-crystallin. The Beta A1- and Beta A3-crystallin proteins are encoded by a single mRNA. They differ by only 17 amino acids, and Beta A1-crystallin is generated by use of an alternate translation initiation site.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CRYBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge