Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PRAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12676R-A680
BOSSBS-12676R-A680EA 890 EUR InStock
BOSSBS-12676R-A680
Anti-PRAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Complete remission of acute promyelocytic leukaemia can be achieved by treating patients with retinoic acid, and PML-RAR-a (promyelocytic leukaemia-retinoic acid receptor alpha fusion protein) plays a major role in mediating retinoic acid effects in leukaemia cells. The retinoic acid-induced gene, PRAM-1 (PML-RAR-a target gene encoding an adaptor molecule 1) encodes an adaptor protein which is expressed and modulated during normal human myelopoiesis. PRAM-1 expression is hindered by expression of PML-RAR-a. The 718 amino acid PRAM-1 protein contains eight N-terminal proline-rich repeats and several proline residues that are clustered as type I or type II SH3 recognition motifs. PRAM-1 demonstrates expression in hematopoietic tissues and lung.

Type: Primary
Antigen: PRAM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 551-650/718
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge