Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SP6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12704R-FITC
BOSSBS-12704R-FITCEA 900 EUR InStock
BOSSBS-12704R-FITC
Anti-SP6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. Sp6, also known as EPFN, EPIPROFIN or KLF14, is a 376 amino acid protein that localizes to the nucleus and contains three C2H2-type zinc fingers. Expressed ubiquitously with higher expression in developing teeth, hair follicles and limb buds, Sp6 functions to bind GC-rich sequences and related GT and CACCC boxes, thereby promoting cellular proliferation. Human Sp6 shares 96% sequence homology with its mouse counterpart, suggesting a conserved role between species. The gene encoding Sp6 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SP6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge