Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PPP1R13B Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1282R-A555
BOSSBS-1282R-A555EA 876 EUR InStock
BOSSBS-1282R-A555
Anti-PPP1R13B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
iASPP (inhibitor of apoptosis stimulating protein of p53, RelA-associated inhibitor (RAI), NFKap B-interacting protein 1, protein phosphatase 1 regulatory subunit 13-like, PPP1R13L) is the third member of the ASPP family of proteins. Unlike ASPP1 and ASPP2, which interact with p53 and enhance its ability to induce apoptosis, iASPP inhibits apoptosis induced by p53 overexpression. iASPP has been identified in a yeast-two hybrid screen as an interacting protein of the p65 subunit of NFKap B (RelA),3 interacts with NFKap B in vivo, and acts as an efficient inhibitor of HIV-1 gene expression in which both NFKap b and Sp1 play major roles. iASPP expression is upregulated in human breast carcinomas expressing wild-type p53, and gene overexpression may play an important role in the leukemogenesis and/or progression of acute leukemia. Alternate Names: apoptosis stimulating protein of P53 1; Apoptosis-stimulating of p53 protein 2; Tumor suppressor p53-binding protein 2; p53-binding protein 2; p53BP2; 53BP2; Bcl2-binding protein; Bbp; Renal carcinoma antigen NY-REN-51.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPP1R13B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge