Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRYBA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12859R-A647
BOSSBS-12859R-A647EA 893 EUR InStock
BOSSBS-12859R-A647
Anti-CRYBA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Crystallins are the major proteins of the vertebrate eye lens, where they maintain the transparency and refractive index of the lens. Crystallins are divided into a, b, and g families, and the b- and g-crystallins also comprise a superfamily. Crystallins usually contain seven distinctive protein regions, including four homologous motifs, a connecting peptide, and N- and C-terminal extensions. b-crystallins constitute the major lens structural proteins. They associate into dimers, tetramers, and higher order aggregates. The b-crystallin subfamily is composed of several gene products, including bA1-, bA2-, bA3-, bA4-, bB1-, bB2- and bB3-crystallin. The bA1- and bA3-crystallin proteins are encoded by a single mRNA. They differ by only 17 amino acids, and bA1-crystallin is generated by use of an alternate translation initiation site. The genes for bA4-, bB1-, bB2- and bB3-crystallin are clustered on human chromosome 22q11, while the genes for bA3/A1- and bA2-crystallin map to human chromosomes 17q11 and 2q34, respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRYBA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge