Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRYGS Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12860R-A350
BOSSBS-12860R-A350EA 893 EUR InStock
BOSSBS-12860R-A350
Anti-CRYGS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Crystallins are separated into two classes:taxon-specific, or enzyme, and ubiquitous. The latter classconstitutes the major proteins of vertebrate eye lens and maintainsthe transparency and refractive index of the lens. Since lenscentral fiber cells lose their nuclei during development, thesecrystallins are made and then retained throughout life, making themextremely stable proteins. Mammalian lens crystallins are dividedinto alpha, beta, and gamma families; beta and gamma crystallinsare also considered as a superfamily. Alpha and beta families arefurther divided into acidic and basic groups. Seven protein regionsexist in crystallins: four homologous motifs, a connecting peptide,and N- and C-terminal extensions. Gamma-crystallins are ahomogeneous group of highly symmetrical, monomeric proteinstypically lacking connecting peptides and terminal extensions. Theyare differentially regulated after early development. This geneencodes a protein initially considered to be a beta-crystallin butthe encoded protein is monomeric and has greater sequencesimilarity to other gamma-crystallins. This gene encodes the mostsignificant gamma-crystallin in adult eye lens tissue. Whether dueto aging or mutations in specific genes, gamma-crystallins havebeen involved in cataract formation. [provided by RefSeq, Jul2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRYGS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge